Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGTAGCGAGCCGGGCTGACAGT[A/G]GAGAAGGTCCGGCCTGGGTAGCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612865 MIM: 606378 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP5KL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIP5KL1 - phosphatidylinositol-4-phosphate 5-kinase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135219.1 | 2215 | Silent Mutation | TCC,TCT | S,S 374 | NP_001128691.1 | |
NM_173492.1 | 2215 | Silent Mutation | TCC,TCT | S,S 171 | NP_775763.1 | |
XM_017014279.1 | 2215 | UTR 3 | XP_016869768.1 |
ST6GALNAC4 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |