Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCAGGCAGGCCCAGTGGGCCAG[A/G]GGCACTCGACACAGTGGGCAGATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF12 - kinesin family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138424.1 | 1616 | Silent Mutation | CCC,CCT | P,P 379 | NP_612433.1 | |
XM_005251683.4 | 1616 | Silent Mutation | CCC,CCT | P,P 517 | XP_005251740.1 | |
XM_006716947.2 | 1616 | Silent Mutation | CCC,CCT | P,P 483 | XP_006717010.1 | |
XM_011518190.2 | 1616 | Silent Mutation | CCC,CCT | P,P 575 | XP_011516492.1 | |
XM_011518191.2 | 1616 | Silent Mutation | CCC,CCT | P,P 575 | XP_011516493.1 | |
XM_011518192.2 | 1616 | Silent Mutation | CCC,CCT | P,P 575 | XP_011516494.1 | |
XM_011518193.2 | 1616 | Intron | XP_011516495.1 | |||
XM_017014240.1 | 1616 | Silent Mutation | CCC,CCT | P,P 252 | XP_016869729.1 |