Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCCGGGCCCTGGTGCCACTCAA[C/G]CTTCCAGCTGGACTTGGGCCTCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602337 | ||||||||||||||||||||
Literature Links: |
ROR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ROR2 - receptor tyrosine kinase like orphan receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318204.1 | 1733 | Intron | NP_001305133.1 | |||
NM_004560.3 | 1733 | Missense Mutation | GCT,GGT | A,G 943 | NP_004551.2 | |
XM_005252008.4 | 1733 | Missense Mutation | GCT,GGT | A,G 803 | XP_005252065.1 | |
XM_005252009.3 | 1733 | Missense Mutation | GCT,GGT | A,G 542 | XP_005252066.1 | |
XM_006717121.3 | 1733 | Missense Mutation | GCT,GGT | A,G 803 | XP_006717184.1 | |
XM_017014762.1 | 1733 | Missense Mutation | GCT,GGT | A,G 940 | XP_016870251.1 | |
XM_017014763.1 | 1733 | Missense Mutation | GCT,GGT | A,G 803 | XP_016870252.1 |