Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATATTTACCCCCAGTTGGTAAT[A/G]CCAAGAGAGGGAAGGTCTTCATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605962 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376271 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105376271 - uncharacterized LOC105376271 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABEPK - Rab9 effector protein with kelch motifs | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174152.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | NP_001167623.1 | |
NM_001174153.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | NP_001167624.1 | |
NM_005833.3 | 462 | Missense Mutation | ACC,GCC | T,A 45 | NP_005824.2 | |
XM_005251640.4 | 462 | Missense Mutation | ACC,GCC | T,A 53 | XP_005251697.1 | |
XM_005251641.4 | 462 | Missense Mutation | ACC,GCC | T,A 53 | XP_005251698.1 | |
XM_005251642.4 | 462 | Missense Mutation | ACC,GCC | T,A 53 | XP_005251699.1 | |
XM_005251644.4 | 462 | Missense Mutation | ACC,GCC | T,A 53 | XP_005251701.1 | |
XM_011518120.2 | 462 | Missense Mutation | ACC,GCC | T,A 53 | XP_011516422.1 | |
XM_017014177.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | XP_016869666.1 | |
XM_017014178.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | XP_016869667.1 | |
XM_017014179.1 | 462 | UTR 5 | XP_016869668.1 | |||
XM_017014180.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | XP_016869669.1 | |
XM_017014181.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | XP_016869670.1 | |
XM_017014182.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | XP_016869671.1 | |
XM_017014183.1 | 462 | Missense Mutation | ACC,GCC | T,A 45 | XP_016869672.1 |