Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCTCCCGCTGCTCCTGCAGCCGG[C/T]GCTGGGCCCCTCTCTTCTCGTCGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609182 MIM: 610160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35D2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35D2 - solute carrier family 35 member D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF367 - zinc finger protein 367 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153695.3 | 1320 | Missense Mutation | CAC,CGC | H,R 322 | NP_710162.1 | |
XM_011518345.2 | 1320 | Intron | XP_011516647.1 |