Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCTTCTGTGTGTTGCAGGGCGC[A/T]GACTGTGGCGCCAAGGTTAGCGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604346 MIM: 612057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN1B1 - mannosidase alpha class 1B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN1B1-AS1 - MAN1B1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAPCD2 - suppressor APC domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UAP1L1 - UDP-N-acetylglucosamine pyrophosphorylase 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207309.2 | 886 | Silent Mutation | GCA,GCT | A,A 276 | NP_997192.2 | |
XM_006717317.3 | 886 | Silent Mutation | GCA,GCT | A,A 276 | XP_006717380.1 | |
XM_011519182.2 | 886 | Silent Mutation | GCA,GCT | A,A 109 | XP_011517484.1 |