Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACAGATGCCCTCTACCAGAAA[T/G]TAGACTTTACAGCACATCAGAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF658 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF658 - zinc finger protein 658 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317916.1 | Intron | NP_001304845.1 | ||||
NM_033160.6 | Intron | NP_149350.3 | ||||
XM_005272515.4 | Intron | XP_005272572.1 | ||||
XM_011545679.2 | Intron | XP_011543981.1 | ||||
XM_017014614.1 | Intron | XP_016870103.1 |