Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTAGGGCCCACGCCCACCATCT[C/T]ACAGTCCAAGGCTAAGGCTCTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604134 MIM: 602930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS13 - ADAM metallopeptidase with thrombospondin type 1 motif 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REXO4 - REX4 homolog, 3'-5' exonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279349.1 | 680 | Intron | NP_001266278.1 | |||
NM_001279350.1 | 680 | Missense Mutation | AAG,GAG | K,E 112 | NP_001266279.1 | |
NM_001279351.1 | 680 | Missense Mutation | AAG,GAG | K,E 156 | NP_001266280.1 | |
NM_020385.3 | 680 | Missense Mutation | AAG,GAG | K,E 249 | NP_065118.2 |
STKLD1 - serine/threonine kinase like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |