Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTACCTGACCTTGGTGTTGAG[C/G]TTCAGGTATGCCACATAGTGTGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B3 - hydroxysteroid 17-beta dehydrogenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000197.1 | 3344 | Missense Mutation | AAC,AAG | N,K 304 | NP_000188.1 | |
XM_011518618.2 | 3344 | Missense Mutation | AAC,AAG | N,K 304 | XP_011516920.1 | |
XM_011518619.2 | 3344 | Missense Mutation | AAC,AAG | N,K 304 | XP_011516921.1 | |
XM_017014671.1 | 3344 | Missense Mutation | AAC,AAG | N,K 304 | XP_016870160.1 | |
XM_017014672.1 | 3344 | Missense Mutation | AAC,AAG | N,K 304 | XP_016870161.1 | |
XM_017014673.1 | 3344 | Missense Mutation | AAC,AAG | N,K 292 | XP_016870162.1 | |
XM_017014674.1 | 3344 | Missense Mutation | AAC,AAG | N,K 268 | XP_016870163.1 | |
XM_017014675.1 | 3344 | Missense Mutation | AAC,AAG | N,K 250 | XP_016870164.1 | |
XM_017014676.1 | 3344 | Intron | XP_016870165.1 | |||
XM_017014677.1 | 3344 | Missense Mutation | AAC,AAG | N,K 184 | XP_016870166.1 |