Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATAGGTCTATTTAAACCTTACCC[A/T]TTAAATTAGGTAGTTCTCCAAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613433 MIM: 612194 MIM: 616292 MIM: 615646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAUS6 - HAUS augmin like complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270890.1 | 1943 | Intron | NP_001257819.1 | |||
NM_017645.4 | 1943 | Intron | NP_060115.3 | |||
XM_011517935.2 | 1943 | Missense Mutation | AAG,ATG | K,M 504 | XP_011516237.1 | |
XM_017014830.1 | 1943 | Intron | XP_016870319.1 |
RRAGA - Ras related GTP binding A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAXO1 - stabilizer of axonemal microtubules 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA8 - small Cajal body-specific RNA 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |