Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGTTCCTCGCTAATGGCCTT[A/G]TACTTCATCTTCTGGGCATAGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603179 MIM: 190990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA9 - carbonic anhydrase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPM2 - tropomyosin 2 (beta) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301226.1 | 1040 | Intron | NP_001288155.1 | |||
NM_001301227.1 | 1040 | Silent Mutation | TAC,TAT | Y,Y 267 | NP_001288156.1 | |
NM_003289.3 | 1040 | Silent Mutation | TAC,TAT | Y,Y 267 | NP_003280.2 | |
NM_213674.1 | 1040 | Intron | NP_998839.1 | |||
XM_017015087.1 | 1040 | UTR 3 | XP_016870576.1 | |||
XM_017015088.1 | 1040 | Intron | XP_016870577.1 | |||
XM_017015089.1 | 1040 | Intron | XP_016870578.1 | |||
XM_017015090.1 | 1040 | Silent Mutation | TAC,TAT | Y,Y 267 | XP_016870579.1 | |
XM_017015091.1 | 1040 | Silent Mutation | TAC,TAT | Y,Y 267 | XP_016870580.1 | |
XM_017015092.1 | 1040 | UTR 3 | XP_016870581.1 | |||
XM_017015093.1 | 1040 | Intron | XP_016870582.1 |