Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCACCAGATGTCTGATTCGTAG[C/T]GGGATCCGAGTTGGCTACACCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRHPR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRHPR - glyoxylate reductase/hydroxypyruvate reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012203.1 | 343 | Silent Mutation | CGC,CGT | R,R 96 | NP_036335.1 | |
XM_005251631.1 | 343 | Intron | XP_005251688.1 | |||
XM_011518073.2 | 343 | UTR 5 | XP_011516375.1 | |||
XM_017015320.1 | 343 | Silent Mutation | CGC,CGT | R,R 96 | XP_016870809.1 | |
XM_017015321.1 | 343 | Silent Mutation | CGC,CGT | R,R 96 | XP_016870810.1 | |
XM_017015322.1 | 343 | Intron | XP_016870811.1 | |||
XM_017015323.1 | 343 | UTR 5 | XP_016870812.1 |