Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGAGGAAAGGATGCGAGTGGAAG[C/G]GCTTCCTGAAGAATAACTGGGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC1A1 - solute carrier family 1 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004170.5 | 241 | Missense Mutation | CGC,GGC | R,G 13 | NP_004161.4 | |
XM_011518007.1 | 241 | Missense Mutation | CGC,GGC | R,G 13 | XP_011516309.1 | |
XM_011518008.2 | 241 | Intron | XP_011516310.1 | |||
XM_011518009.2 | 241 | Intron | XP_011516311.1 | |||
XM_011518010.1 | 241 | Missense Mutation | CGC,GGC | R,G 13 | XP_011516312.1 | |
XM_017015042.1 | 241 | Missense Mutation | CGC,GGC | R,G 13 | XP_016870531.1 | |
XM_017015043.1 | 241 | Missense Mutation | CGC,GGC | R,G 13 | XP_016870532.1 |