Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCGCAAGGCCTCCTGGAAAATG[C/T]GGGGGTCCGGCTTGGGCCAGCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BSPRY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BSPRY - B-box and SPRY domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HDHD3 - haloacid dehalogenase like hydrolase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304509.1 | 1083 | Missense Mutation | CAC,CGC | H,R 171 | NP_001291438.1 | |
NM_001304510.1 | 1083 | Missense Mutation | CAC,CGC | H,R 171 | NP_001291439.1 | |
NM_001304511.1 | 1083 | Missense Mutation | CAC,CGC | H,R 171 | NP_001291440.1 | |
NM_031219.3 | 1083 | Missense Mutation | CAC,CGC | H,R 171 | NP_112496.1 |