Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAGCAACAAATTCTGACATCAT[A/G]TTTATGGTTTCTGTATCTTTGTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179730 MIM: 179740 | ||||||||||||||||||||
Literature Links: |
RLN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RLN1 - relaxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006911.3 | 387 | Intron | NP_008842.1 |
RLN2 - relaxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005059.3 | 387 | UTR 3 | NP_005050.2 | |||
NM_134441.2 | 387 | Silent Mutation | AAC,AAT | N,N 85 | NP_604390.1 | |
XM_017015011.1 | 387 | Silent Mutation | AAC,AAT | N,N 69 | XP_016870500.1 |