Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCTTGCTGCTGCTCAGCTGCC[C/T]GCTGACATCCTGCAGACGCTTTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601541 | ||||||||||||||||||||
Literature Links: |
BRD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRD3 - bromodomain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007371.3 | 2568 | Missense Mutation | AGG,GGG | R,G 677 | NP_031397.1 | |
XM_006717291.2 | 2568 | Missense Mutation | AGG,GGG | R,G 677 | XP_006717354.1 | |
XM_011519052.2 | 2568 | Missense Mutation | AGG,GGG | R,G 677 | XP_011517354.1 | |
XM_017015165.1 | 2568 | Missense Mutation | AGG,GGG | R,G 676 | XP_016870654.1 | |
XM_017015166.1 | 2568 | Missense Mutation | AGG,GGG | R,G 356 | XP_016870655.1 |
LINC00094 - long intergenic non-protein coding RNA 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |