Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTTTTTTAACCACTGTGCACC[C/T]GATAACAGGTTGGAGATCTCCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf153 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf153 - chromosome 9 open reading frame 153 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276366.1 | 375 | Missense Mutation | AGG,GGG | R,G 81 | NP_001263295.1 | |
NM_001276367.1 | 375 | Missense Mutation | AGG,GGG | R,G 81 | NP_001263296.1 | |
NM_001276368.1 | 375 | Missense Mutation | AGG,GGG | R,G 81 | NP_001263297.1 |