Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGGAGGATCTCCTCGCGGTCCCGCC[A/G]GCTGCGCCACGCCTCCAATGCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604722 MIM: 608052 | ||||||||||||||||||||
Literature Links: |
SH2D3C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH2D3C - SH2 domain containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOR2A - torsin family 2 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085347.2 | 760 | Missense Mutation | CGG,TGG | R,W 216 | NP_001078816.1 | |
NM_001134430.2 | 760 | Intron | NP_001127902.1 | |||
NM_001134431.2 | 760 | UTR 3 | NP_001127903.1 | |||
NM_001252018.1 | 760 | Missense Mutation | CGG,TGG | R,W 54 | NP_001238947.1 | |
NM_001252021.1 | 760 | Missense Mutation | CGG,TGG | R,W 54 | NP_001238950.1 | |
NM_001252023.1 | 760 | Intron | NP_001238952.1 | |||
NM_130459.3 | 760 | UTR 3 | NP_569726.2 | |||
XM_011518554.1 | 760 | Missense Mutation | CGG,TGG | R,W 215 | XP_011516856.1 |
TTC16 - tetratricopeptide repeat domain 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |