Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGCTCGTGTGGGTGCCCATGG[A/G]CATCTGGCCAAAGAGGTTGGGCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609720 MIM: 613633 | ||||||||||||||||||||
Literature Links: |
CRB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRB2 - crumbs 2, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DENND1A - DENN domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020946.1 | 3808 | Missense Mutation | CCC,TCC | P,S 900 | NP_065997.1 | |
NM_024820.2 | 3808 | Intron | NP_079096.2 | |||
XM_005252109.2 | 3808 | Missense Mutation | CCC,TCC | P,S 961 | XP_005252166.1 | |
XM_005252111.4 | 3808 | Missense Mutation | CCC,TCC | P,S 915 | XP_005252168.1 | |
XM_005252113.4 | 3808 | Intron | XP_005252170.1 | |||
XM_006717195.3 | 3808 | Missense Mutation | CCC,TCC | P,S 958 | XP_006717258.1 | |
XM_011518882.2 | 3808 | Missense Mutation | CCC,TCC | P,S 976 | XP_011517184.1 | |
XM_011518883.2 | 3808 | Missense Mutation | CCC,TCC | P,S 957 | XP_011517185.1 | |
XM_011518884.2 | 3808 | Missense Mutation | CCC,TCC | P,S 947 | XP_011517186.1 | |
XM_011518885.2 | 3808 | Missense Mutation | CCC,TCC | P,S 940 | XP_011517187.1 | |
XM_011518886.2 | 3808 | Missense Mutation | CCC,TCC | P,S 933 | XP_011517188.1 | |
XM_011518887.2 | 3808 | Intron | XP_011517189.1 | |||
XM_017014947.1 | 3808 | Missense Mutation | CCC,TCC | P,S 949 | XP_016870436.1 | |
XM_017014948.1 | 3808 | Missense Mutation | CCC,TCC | P,S 943 | XP_016870437.1 | |
XM_017014949.1 | 3808 | Missense Mutation | CCC,TCC | P,S 934 | XP_016870438.1 | |
XM_017014950.1 | 3808 | Missense Mutation | CCC,TCC | P,S 916 | XP_016870439.1 | |
XM_017014951.1 | 3808 | Missense Mutation | CCC,TCC | P,S 911 | XP_016870440.1 | |
XM_017014952.1 | 3808 | Missense Mutation | CCC,TCC | P,S 702 | XP_016870441.1 |