Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAGCCGGCCCTTCTCAAAGTAGT[G/T]GTAAGGCACTGAGGGGTGGCTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606378 MIM: 610135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST6GALNAC4 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175039.3 | 1147 | Missense Mutation | AAC,CAC | N,H 250 | NP_778204.1 | |
NM_175040.3 | 1147 | Missense Mutation | AAC,CAC | N,H 166 | NP_778205.1 | |
XM_011518527.2 | 1147 | Missense Mutation | AAC,CAC | N,H 273 | XP_011516829.1 | |
XM_011518529.2 | 1147 | UTR 3 | XP_011516831.1 |
ST6GALNAC6 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |