Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCCCCTACAACCAGCTGTTCAT[C/T]AACAATGAATGGCAAGATGCAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100670 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH1B1 - aldehyde dehydrogenase 1 family member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000692.4 | 276 | Silent Mutation | ATC,ATT | I,I 41 | NP_000683.3 | |
XM_011517802.2 | 276 | Silent Mutation | ATC,ATT | I,I 41 | XP_011516104.1 |