Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAACAACACAAACGTTCATCTCA[A/G]GCTGGTTAACCCTCTGCGACATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604303 MIM: 604304 MIM: 603722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTL7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTL7A - actin like 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006687.3 | 484 | Missense Mutation | AAG,AGG | K,R 130 | NP_006678.1 |
ACTL7B - actin like 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IKBKAP - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |