Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTTTTTATCTGGTGGCTTAAA[G/T]TAGGAATAAACAGGCGCTGCTGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RBM18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM18 - RNA binding motif protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033117.3 | 673 | Nonsense Mutation | TAA,TAC | *,Y 169 | NP_149108.1 | |
XM_006717319.1 | 673 | Nonsense Mutation | TAA,TAC | *,Y 169 | XP_006717382.1 |