Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGATGAGGCTTCCAGTGGCTC[C/G]CAAGCCTCTTAGCGAGAGACTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602238 | ||||||||||||||||||||
Literature Links: |
EXOSC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOSC2 - exosome component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282708.1 | 64 | Missense Mutation | CCC,CGC | P,R 11 | NP_001269637.1 | |
NM_001282709.1 | 64 | Missense Mutation | CCC,CGC | P,R 11 | NP_001269638.1 | |
NM_014285.6 | 64 | Missense Mutation | CCC,CGC | P,R 11 | NP_055100.2 | |
XM_005272176.2 | 64 | UTR 5 | XP_005272233.1 | |||
XM_006717023.2 | 64 | UTR 5 | XP_006717086.1 | |||
XM_017014558.1 | 64 | UTR 5 | XP_016870047.1 |