Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCATCAGGTTCATCAGTATCAA[A/G]TGCTGAAAGCTCTAACTCGATATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RC3H2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RC3H2 - ring finger and CCCH-type domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100588.1 | 3431 | Missense Mutation | CTT,TTT | L,F 1064 | NP_001094058.1 | |
NM_018835.2 | 3431 | Intron | NP_061323.2 |