Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606946 MIM: 138249 MIM: 613505 | ||||||||||||||||||||
Literature Links: |
ANAPC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANAPC2 - anaphase promoting complex subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRIN1 - glutamate ionotropic receptor NMDA type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000832.6 | 859 | Intron | NP_000823.4 | |||
NM_001185090.1 | 859 | Intron | NP_001172019.1 | |||
NM_001185091.1 | 859 | Intron | NP_001172020.1 | |||
NM_007327.3 | 859 | Intron | NP_015566.1 | |||
NM_021569.3 | 859 | Intron | NP_067544.1 | |||
XM_005266071.3 | 859 | Intron | XP_005266128.1 | |||
XM_005266072.3 | 859 | Intron | XP_005266129.1 | |||
XM_005266073.4 | 859 | Intron | XP_005266130.1 | |||
XM_011518583.2 | 859 | Intron | XP_011516885.1 |
LRRC26 - leucine rich repeat containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013653.2 | 859 | Nonsense Mutation | CAG,TAG | Q,* 255 | NP_001013675.1 |
MIR3621 - microRNA 3621 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM210 - transmembrane protein 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |