Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTGTCATCCTTGTCAGTTCTC[C/T]TTTTCACTTCTCTGAGACAGAGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611344 | ||||||||||||||||||||
Literature Links: |
RASEF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RASEF - RAS and EF-hand domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152573.3 | 2370 | Missense Mutation | AAG,AGG | K,R 710 | NP_689786.2 | |
XM_005251730.3 | 2370 | Missense Mutation | AAG,AGG | K,R 687 | XP_005251787.1 | |
XM_005251731.3 | 2370 | Missense Mutation | AAG,AGG | K,R 581 | XP_005251788.1 |