Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTGACCGCCTGGTGCAGGACA[C/T]GCCTTTCCTGACAGGCCATGGGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CARD19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CARD19 - caspase recruitment domain family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318010.1 | 264 | Missense Mutation | ACG,ATG | T,M 14 | NP_001304939.1 | |
NM_001318011.1 | 264 | Missense Mutation | ACG,ATG | T,M 14 | NP_001304940.1 | |
NM_032310.4 | 264 | Missense Mutation | ACG,ATG | T,M 14 | NP_115686.3 |