Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGACCAGAAAGCTGGGAGCCCTG[C/T]GTGGAGGTATGTGGCTGGAGTCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603030 | ||||||||||||||||||||
Literature Links: |
TLR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TLR4 - toll like receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003266.3 | 385 | UTR 5 | NP_003257.1 | |||
NM_138554.4 | 385 | Silent Mutation | TGC,TGT | C,C 29 | NP_612564.1 | |
NM_138557.2 | 385 | UTR 5 | NP_612567.1 |