Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAATCTGTCCAAGAGCTGGCTT[C/G]ATGTTCAGGTCTCCAACACCTATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPD52L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TPD52L3 - tumor protein D52-like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001874.2 | 491 | Missense Mutation | CAT,GAT | H,D 82 | NP_001001874.1 | |
NM_001001875.3 | 491 | Missense Mutation | CAT,GAT | H,D 82 | NP_001001875.1 | |
NM_033516.5 | 491 | Missense Mutation | CAT,GAT | H,D 82 | NP_277051.3 | |
XM_017015280.1 | 491 | Missense Mutation | CAT,GAT | H,D 82 | XP_016870769.1 |