Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCTCATCCTCATCCTCATCTTC[A/C]TCATCCTCATCCTCATCATCATCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300314 | ||||||||||||||||||||
Literature Links: |
TAF7L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAF7L - TATA-box binding protein associated factor 7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168474.1 | 878 | Missense Mutation | GAG,GAT | E,D 254 | NP_001161946.1 | |
NM_024885.3 | 878 | Missense Mutation | GAG,GAT | E,D 340 | NP_079161.3 | |
XM_005262152.1 | 878 | Missense Mutation | GAG,GAT | E,D 255 | XP_005262209.1 | |
XM_006724664.1 | 878 | Missense Mutation | GAG,GAT | E,D 341 | XP_006724727.1 |