Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTCTGCTGCCTGGGCACTGGC[C/T]CCTGATGGCGCTGCCAGGGGCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300283 MIM: 300005 MIM: 300929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRAK1 - interleukin 1 receptor associated kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025242.1 | 1718 | Silent Mutation | GGA,GGG | G,G 539 | NP_001020413.1 | |
NM_001025243.1 | 1718 | Silent Mutation | GGA,GGG | G,G 490 | NP_001020414.1 | |
NM_001569.3 | 1718 | Silent Mutation | GGA,GGG | G,G 569 | NP_001560.2 | |
XM_005274668.3 | 1718 | Silent Mutation | GGA,GGG | G,G 565 | XP_005274725.1 | |
XM_011531158.2 | 1718 | Silent Mutation | GGA,GGG | G,G 460 | XP_011529460.1 |
MECP2 - methyl-CpG binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR718 - microRNA 718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |