Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGTCAGCGGGCCCGTCGGGGGT[G/T]CTGCGATCCTGGATCACGTTAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NHSL2 - NHS like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013627.2 | 697 | Intron | NP_001013649.2 | |||
XM_011530932.1 | 697 | Intron | XP_011529234.1 | |||
XM_011530933.1 | 697 | Intron | XP_011529235.1 | |||
XM_011530934.2 | 697 | Intron | XP_011529236.1 | |||
XM_017029473.1 | 697 | Intron | XP_016884962.1 | |||
XM_017029474.1 | 697 | Intron | XP_016884963.1 | |||
XM_017029475.1 | 697 | Intron | XP_016884964.1 | |||
XM_017029476.1 | 697 | Intron | XP_016884965.1 | |||
XM_017029477.1 | 697 | Intron | XP_016884966.1 |
RGAG4 - retrotransposon gag domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024455.3 | 697 | Missense Mutation | AGA,AGC | R,S 112 | NP_001019626.1 |