Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATCCTCAGTGGACACAAAATTG[A/T]CATTATCAGAATCATCATTATATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300553 | ||||||||||||||||||||
Literature Links: |
BRWD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRWD3 - bromodomain and WD repeat domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153252.4 | 5560 | Missense Mutation | GAC,GTC | D,V 1766 | NP_694984.4 | |
XM_005262113.3 | 5560 | Missense Mutation | GAC,GTC | D,V 1716 | XP_005262170.1 | |
XM_017029384.1 | 5560 | Missense Mutation | GAC,GTC | D,V 1362 | XP_016884873.1 | |
XM_017029385.1 | 5560 | Intron | XP_016884874.1 |