Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCATAGCAGCAGCCTCCGAAG[C/G]ACCTGCCCCCACCCTCAGGTAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRP2 - dystrophin related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171184.1 | 626 | Intron | NP_001164655.1 | |||
NM_001939.2 | 626 | Silent Mutation | AGC,AGG | S,R 33 | NP_001930.2 | |
XM_017029333.1 | 626 | Silent Mutation | AGC,AGG | S,R 33 | XP_016884822.1 | |
XM_017029334.1 | 626 | UTR 5 | XP_016884823.1 |