Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCAGGGATGGCCATCAGAAGAG[C/G]TGCAGGCCAGTTTTGAGCCCATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300964 MIM: 300889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAS1L - LAS1 like, ribosome biogenesis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170649.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 715 | NP_001164120.1 | |
NM_001170650.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 673 | NP_001164121.1 | |
NM_031206.4 | 2218 | Missense Mutation | CAC,CAG | H,Q 732 | NP_112483.1 | |
XM_005262301.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 731 | XP_005262358.1 | |
XM_005262304.1 | 2218 | Intron | XP_005262361.1 | |||
XM_005262305.3 | 2218 | Intron | XP_005262362.1 | |||
XM_005262306.3 | 2218 | Intron | XP_005262363.1 | |||
XM_011531045.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 690 | XP_011529347.1 | |
XM_011531046.1 | 2218 | Intron | XP_011529348.1 | |||
XM_017029877.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 714 | XP_016885366.1 | |
XM_017029878.1 | 2218 | Intron | XP_016885367.1 | |||
XM_017029879.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 430 | XP_016885368.1 | |
XM_017029880.1 | 2218 | Missense Mutation | CAC,CAG | H,Q 413 | XP_016885369.1 |
ZC3H12B - zinc finger CCCH-type containing 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |