Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCTGTCGTCACTGCCCAGGGG[C/T]TTGATGTCCCCGTTGAGCGATGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 308840 | ||||||||||||||||||||
Literature Links: |
L1CAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
L1CAM - L1 cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000425.4 | 3818 | Silent Mutation | AAA,AAG | K,K 1200 | NP_000416.1 | |
NM_001143963.2 | 3818 | Silent Mutation | AAA,AAG | K,K 1191 | NP_001137435.1 | |
NM_001278116.1 | 3818 | Silent Mutation | AAA,AAG | K,K 1200 | NP_001265045.1 | |
NM_024003.3 | 3818 | Silent Mutation | AAA,AAG | K,K 1196 | NP_076493.1 |