Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGGCCTGGCCTGTGGCTGATA[C/T]CTTCTACCGTATCCACCGAAGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300521 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDPD2 - glycerophosphodiester phosphodiesterase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171191.1 | 410 | Missense Mutation | ACC,ATC | T,I 99 | NP_001164662.1 | |
NM_001171192.1 | 410 | Missense Mutation | ACC,ATC | T,I 178 | NP_001164663.1 | |
NM_001171193.1 | 410 | Missense Mutation | ACC,ATC | T,I 99 | NP_001164664.1 | |
NM_017711.3 | 410 | Missense Mutation | ACC,ATC | T,I 178 | NP_060181.2 | |
XM_011530977.1 | 410 | Missense Mutation | ACC,ATC | T,I 51 | XP_011529279.1 | |
XM_017029614.1 | 410 | UTR 5 | XP_016885103.1 |
KIF4A - kinesin family member 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105373244 - uncharacterized LOC105373244 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011531081.2 | 410 | Intron | XP_011529383.1 | |||
XM_011531082.2 | 410 | Intron | XP_011529384.1 |