Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTGGACGCACTTTGCTTGTGA[A/G]GCCAGAATATATGCTCATGTGATTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300628 MIM: 300547 | ||||||||||||||||||||
Literature Links: |
FRMD7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRMD7 - FERM domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306193.1 | 1527 | Missense Mutation | CTC,TTC | L,F 448 | NP_001293122.1 | |
NM_194277.2 | 1527 | Missense Mutation | CTC,TTC | L,F 463 | NP_919253.1 | |
XM_017029947.1 | 1527 | Missense Mutation | CTC,TTC | L,F 447 | XP_016885436.1 | |
XM_017029948.1 | 1527 | Missense Mutation | CTC,TTC | L,F 378 | XP_016885437.1 | |
XM_017029949.1 | 1527 | Missense Mutation | CTC,TTC | L,F 305 | XP_016885438.1 |
STK26 - serine/threonine protein kinase 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |