Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAAAATGGGCTGGAGGCTCGTG[G/A]TTTCCTGGACATCTTCACCAGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300944 MIM: 300177 MIM: 300549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSAG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSAG1 - chondrosarcoma associated gene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102576.2 | 447 | Intron | NP_001096046.2 | |||
NM_153478.2 | 447 | Missense Mutation | ACA,GCA | T,A 40 | NP_705611.2 |
CSAG4 - CSAG family member 4 (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEA12 - MAGE family member A12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEA2B - MAGE family member A2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |