Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCATAGCCCTTCTCATTGCGGGC[A/G]GCGATGCGGAAGATGATGGCGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300019 MIM: 312420 | ||||||||||||||||||||
Literature Links: |
HCFC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCFC1 - host cell factor C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005334.2 | 6954 | Silent Mutation | GCC,GCT | A,A 1984 | NP_005325.2 | |
XM_006724815.2 | 6954 | Silent Mutation | GCC,GCT | A,A 2029 | XP_006724878.1 | |
XM_006724816.2 | 6954 | Silent Mutation | GCC,GCT | A,A 2028 | XP_006724879.1 | |
XM_011531147.2 | 6954 | Silent Mutation | GCC,GCT | A,A 1988 | XP_011529449.1 | |
XM_011531148.2 | 6954 | Silent Mutation | GCC,GCT | A,A 1987 | XP_011529450.1 | |
XM_017029471.1 | 6954 | Silent Mutation | GCC,GCT | A,A 1962 | XP_016884960.1 | |
XM_017029472.1 | 6954 | Silent Mutation | GCC,GCT | A,A 1725 | XP_016884961.1 |
RENBP - renin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |