Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCTCTCCAGGCAAATTGCTGTG[A/G]AGAAAGTTCCAGATTCCGAGATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300230 | ||||||||||||||||||||
Literature Links: |
CA5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA5B - carbonic anhydrase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007220.3 | 177 | Nonsense Mutation | TGA,TGG | *,W 20 | NP_009151.1 | |
XM_005274442.4 | 177 | Nonsense Mutation | TGA,TGG | *,W 20 | XP_005274499.1 | |
XM_017029247.1 | 177 | Intron | XP_016884736.1 | |||
XM_017029248.1 | 177 | Intron | XP_016884737.1 | |||
XM_017029249.1 | 177 | Intron | XP_016884738.1 |