Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGCCGAAGGGCTGCCCCAGCA[C/G]CAGCAGCTGCCGCAGCTGGAGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300292 | ||||||||||||||||||||
Literature Links: |
FOXP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXP3 - forkhead box P3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R3F - protein phosphatase 1 regulatory subunit 3F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184745.1 | 933 | Intron | NP_001171674.1 | |||
NM_033215.4 | 933 | Missense Mutation | CAC,CAG | H,Q 302 | NP_149992.3 | |
XM_005272687.3 | 933 | Missense Mutation | CAC,CAG | H,Q 302 | XP_005272744.1 | |
XM_017029932.1 | 933 | Intron | XP_016885421.1 | |||
XM_017029933.1 | 933 | Intron | XP_016885422.1 | |||
XM_017029934.1 | 933 | Missense Mutation | CAC,CAG | H,Q 302 | XP_016885423.1 | |
XM_017029935.1 | 933 | Missense Mutation | CAC,CAG | H,Q 302 | XP_016885424.1 |