Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACTGCATTAGAGGGTAAAACTC[A/G]ACTCCGAAAACTTATAGTCCTGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300862 | ||||||||||||||||||||
Literature Links: |
ACOT9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACOT9 - acyl-CoA thioesterase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033583.2 | 1008 | Nonsense Mutation | CGA,TGA | R,* 279 | NP_001028755.2 | |
NM_001037171.1 | 1008 | Nonsense Mutation | CGA,TGA | R,* 288 | NP_001032248.1 | |
XM_005274472.2 | 1008 | Nonsense Mutation | CGA,TGA | R,* 219 | XP_005274529.1 |