Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCATCACATCACTCTTGAGTAGG[A/T]ACAGAGCCTTCCCATCAATTTCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCML2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCML2 - sex comb on midleg-like 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006089.2 | 1966 | Missense Mutation | TAC,TTC | Y,F 666 | NP_006080.1 | |
XM_006724458.1 | 1966 | UTR 3 | XP_006724521.1 | |||
XM_017029219.1 | 1966 | Missense Mutation | TAC,TTC | Y,F 666 | XP_016884708.1 | |
XM_017029220.1 | 1966 | Missense Mutation | TAC,TTC | Y,F 666 | XP_016884709.1 | |
XM_017029221.1 | 1966 | UTR 3 | XP_016884710.1 | |||
XM_017029222.1 | 1966 | Missense Mutation | TAC,TTC | Y,F 402 | XP_016884711.1 |