Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGGATGATGTACCCAGGAAAGC[C/T]AAAAGAAAAACTAATAAGGGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TCEAL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCEAL4 - transcription elongation factor A like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006935.2 | 672 | Silent Mutation | GCC,GCT | A,A 126 | NP_001006936.1 | |
NM_001006937.2 | 672 | Silent Mutation | GCC,GCT | A,A 126 | NP_001006938.1 | |
NM_001300901.1 | 672 | Silent Mutation | GCC,GCT | A,A 269 | NP_001287830.1 | |
NM_001305840.1 | 672 | Silent Mutation | GCC,GCT | A,A 126 | NP_001292769.1 | |
NM_001305841.1 | 672 | Silent Mutation | GCC,GCT | A,A 126 | NP_001292770.1 | |
NM_001305842.1 | 672 | Silent Mutation | GCC,GCT | A,A 126 | NP_001292771.1 | |
NM_024863.5 | 672 | Silent Mutation | GCC,GCT | A,A 126 | NP_079139.4 | |
XM_017029856.1 | 672 | Silent Mutation | GCC,GCT | A,A 188 | XP_016885345.1 | |
XM_017029857.1 | 672 | Silent Mutation | GCC,GCT | A,A 126 | XP_016885346.1 |