Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCCCTTACTTGAGCTGGGGGTG[C/T]GGAGGTCCCCCTGCAGCGGCGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311010 MIM: 313440 MIM: 305370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARAF - A-Raf proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYN1 - synapsin I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006950.3 | 2097 | Silent Mutation | CCA,CCG | P,P 656 | NP_008881.2 | |
NM_133499.2 | 2097 | Silent Mutation | CCA,CCG | P,P 656 | NP_598006.1 |
TIMP1 - TIMP metallopeptidase inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |