Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGATTTCCTTCCACTAACTCTGG[C/T]TGTCTTTTTTGTTGATGCATTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300674 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOSPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOSPD1 - motile sperm domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306188.1 | 478 | Silent Mutation | CAA,CAG | Q,Q 7 | NP_001293117.1 | |
NM_019556.2 | 478 | Silent Mutation | CAA,CAG | Q,Q 7 | NP_062456.1 | |
XM_011531366.2 | 478 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_011529668.1 | |
XM_017029653.1 | 478 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_016885142.1 | |
XM_017029654.1 | 478 | Silent Mutation | CAA,CAG | Q,Q 7 | XP_016885143.1 |