Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGGAGTCAGCCTGGCTGCCGGC[A/T]TGTTCTTACTGGCCCTGAGTCTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 303630 MIM: 303631 | ||||||||||||||||||||
Literature Links: |
COL4A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL4A5 - collagen type IV alpha 5 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000495.4 | 316 | Missense Mutation | ATG,TTG | M,L 12 | NP_000486.1 | |
NM_033380.2 | 316 | Missense Mutation | ATG,TTG | M,L 12 | NP_203699.1 | |
XM_011530849.2 | 316 | Intron | XP_011529151.2 | |||
XM_017029259.1 | 316 | Intron | XP_016884748.1 | |||
XM_017029260.1 | 316 | Intron | XP_016884749.1 | |||
XM_017029261.1 | 316 | Intron | XP_016884750.1 | |||
XM_017029262.1 | 316 | Intron | XP_016884751.1 | |||
XM_017029263.1 | 316 | Intron | XP_016884752.1 |
COL4A6 - collagen type IV alpha 6 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287758.1 | 316 | Intron | NP_001274687.1 | |||
NM_001287759.1 | 316 | Intron | NP_001274688.1 | |||
NM_001287760.1 | 316 | Intron | NP_001274689.1 | |||
NM_001847.3 | 316 | Intron | NP_001838.2 | |||
NM_033641.3 | 316 | Intron | NP_378667.1 | |||
XM_006724617.3 | 316 | Intron | XP_006724680.1 | |||
XM_011530852.2 | 316 | Intron | XP_011529154.1 | |||
XM_011530853.2 | 316 | Intron | XP_011529155.1 | |||
XM_011530854.2 | 316 | Intron | XP_011529156.1 |