Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTCTGGCATTTTAAATTGTTC[C/T]GATTTTGGCAGAATCTTCCCCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XAGE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XAGE3 - X antigen family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130776.1 | 408 | Intron | NP_570132.1 | |||
NM_133179.2 | 408 | Silent Mutation | TCA,TCG | S,S 96 | NP_573440.1 | |
XM_011530768.2 | 408 | Silent Mutation | TCA,TCG | S,S 96 | XP_011529070.1 | |
XM_011530769.2 | 408 | Silent Mutation | TCA,TCG | S,S 96 | XP_011529071.1 | |
XM_011530770.2 | 408 | Silent Mutation | TCA,TCG | S,S 96 | XP_011529072.1 |